EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Plasma cholinesterase activity in the house finch, Carpodacus mexicanus, and the house sparrow, Passer domesticus, collected from pesticide-exposed agricultural land.
PMID 23114840
Abstract
Agricultural lands are exposed to pesticides continuously, which impact on habitat of wild species, such birds. The purpose of this study was to determine the plasma cholinesterase activity in two passeriform wild birds, the house finch (Carpodacus mexicanus) and the house sparrow (Passer domesticus), from agricultural land with exposure to pesticides. No significant difference was found in the house finch between specimens captured in non-agricultural and agricultural lands. However, house sparrows had significant differences in plasma cholinesterase activity, which indicates that this species is more sensitive to enzymatic activity inhibition by pesticides, and they could be indicators of pesticide pollution in agricultural areas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4186
Butyrylcholinesterase human, vial of ≥4 units