EMAIL THIS PAGE TO A FRIEND
Clinical nephrology
In a well presented report of bilateral renal pelvic thrombus formation with subsequent bilateral ureteral obstruction. Introduction.
PMID 23115797
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46036
Brodifacoum, PESTANAL®, analytical standard
C31H23BrO3