EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Capillary electrophoresis to quantitate gossypol enantiomers in cotton flower petals and seed.
PMID 23122406
Abstract
Gossypol is a toxic compound that occurs as a mixture of enantiomers in cotton plant tissues including seed and flower petals. The (-)-enantiomer is more toxic to non-ruminant animals. Efforts to breed cottonseed with a low percentage of (-)-gossypol requires determination of the (+)- to (-)-gossypol ratio in seed and flower petals. We report a method to quantitatively determine the total gossypol and percent of its enantiomers in cotton tissues using high performance capillary electrophoresis (HPCE). The method utilizes a borate buffer at pH 9.3 using a capillary with internal diameter of 50μm, effective length of 24.5cm, 15kV and cassette temperature of 15°C. This method provides high accuracy and reproducible results with a limit of detection of the individual enantiomers of less than 36ng/mL providing base line separation in less than 6min.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8761
(±)-Gossypol from cotton seeds, ≥95% (HPLC)
C30H30O8