EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Isocytosine-based inhibitors of xanthine oxidase: design, synthesis, SAR, PK and in vivo efficacy in rat model of hyperuricemia.
PMID 23122864
Abstract
Structure-activity relationship studies were carried out for lead generation following structure-guided design approach from an isocytosine scaffold identified earlier for xanthine oxidase inhibition. A 470-fold improvement in in vitro IC(50) was obtained in the process. Five most potent compounds with nanomolar IC(50) values were selected for pharmacokinetics and in vivo experiments. The best compound showed good in vivo activity when administered intraperitoneally but was not active by oral route. The results suggest that improvement in oral exposure could improve the in vivo efficacy of this series.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2127
Isocytosine, ≥99%
C4H5N3O