EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Enzymatic assessment of paraoxonase 1 activity on HDL subclasses: a practical zymogram method to assess HDL function.
PMID 23122967
Abstract
We developed a practical method for analysis of PON-1 enzymatic activity in HDL subclasses. The assay uses 4-12% polyacrylamide gradient gels, phenylacetate as substrate coupled with densitometric phenol detection using 4-aminoantipyrine. The measurement PON-1 activity in situ across the HDL subclasses has a strong correlation with the kinetic microplate assay for total PON-1 activity, r=0.91, p<0.001. The same HDL-C level, healthy subjects (n=33) display a large difference in the ratio of PON-1 activity in small vs. large HDL. Since PON-1 activity is larger in HDL(3) we propose that this difference has a potent predictive value for clinical risk assessment and therapeutic choice. Our method also offers the advantage of assessing the distribution of up to six different HDL apolipoproteins in the same gel after transfer. We seek to further dissect the cause of a different distribution of PON-1 activity in HDL subclasses by employing this method that permits practical, inexpensive analysis of antioxidant function of HDL subclasses and has the potential for application in clinical chemistry and to shed some light on the importance of PON-1 distribution.