Phytomedicine : international journal of phytotherapy and phytopharmacology
β-Asarone, an active principle of Acorus calamus rhizome, inhibits morphogenesis, biofilm formation and ergosterol biosynthesis in Candida albicans.
PMID 23123225
Anti-Candida potential of Acorus calamus rhizome and its active principle, β-asarone, was evaluated against the human fungal pathogen, Candida albicans. β-Asarone exhibited promising growth inhibitory activity at 0.5mg/ml and it was fungicidal at 8 mg/ml. Time dependant kill curve assay showed that MFC of β-asarone was highly toxic to C. albicans, killing 99.9% inoculum within 120 min of exposure. β-Asarone caused significant inhibition of C. albicans morphogenesis and biofilm development at sub-inhibitory concentrations. Our data indicate that the growth inhibitory activity of β-asarone might be through inhibition of ergosterol biosynthesis. Hemolytic assay showed that β-asarone is non-toxic, even at concentrations approaching MIC value. Our results suggest that β-asarone may be safe as a topical antifungal agent.
Related Materials