EMAIL THIS PAGE TO A FRIEND
Plant physiology and biochemistry : PPB
Acetohydroxyacid synthase (AHAS) in vivo assay for screening imidazolinone-resistance in sunflower (Helianthus annuus L.).
PMID 23123550
Abstract
The objective of this work was to evaluate the in vivo acetohydroxyacid synthase (AHAS) activity response to imidazolinones and its possible use as a selection method for evaluating AHAS inhibitor resistance. In vivo AHAS assay and the comparison of parameters from dose-response curves have been used as a valid tool for comparing sunflower lines and hybrids differing in imidazolinone resistance. The sunflower resistant genotypes evaluated here were 100-fold and 20-fold more resistant compared with the susceptible line for imazethapyr and imazapyr, respectively. This assay also allowed discrimination of homozygous from heterozygous genotypes for I(mr1) locus that codify for the catalytic subunit of AHAS. The in vivo AHAS assay described in this study was useful for the selection of sunflower genotypes differing in herbicide resistance and could be a useful tool when breeding for imidazolinone resistance in sunflower.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37877
Imazapyr, PESTANAL®, analytical standard
C13H15N3O3
37923
Imazethapyr, PESTANAL®, analytical standard
C15H19N3O3