EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Encapsulation of yeast displaying glucose oxidase on their surface in graphene oxide hydrogel scaffolding and its bioactivation.
PMID 23123800
Abstract
Yeast displaying glucose oxidase on their surface were encapsulated in a graphene oxide hydrogel. The ability of the modified yeast to reduce graphene oxide by glucose assimilation while maintaining viability was tested with time and deemed suitable for biofuel cell applications.