EMAIL THIS PAGE TO A FRIEND
Mitochondrion
Exploiting endobiotic metabolic pathways to target xenobiotic antioxidants to mitochondria.
PMID 23123918
Abstract
Oxidative stress plays a role in a range of human disease entities. Hence, strategies to target antioxidants to mitochondria are an active area of investigation. Triphenylphosphonium cation-based antioxidants and SS-peptides have been described and show significant uptake by mitochondria and effectiveness in animal models of conditions linked to oxidative stress. We tested the hypothesis that the mitochondrial β-oxidation pathway could be exploited to activate the antioxidant phenolic and methimazole prodrugs. Most compounds studied underwent mitochondrial biotransformation to release their antioxidant moieties, and some were cytoprotective in a hypoxia-reoxygenation model in rat cardiomyocytes. These results demonstrate the feasibility of exploiting mitochondrial bioactivation reactions for targeted drug delivery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46429
Methimazole, VETRANAL, analytical standard
C4H6N2S