EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Tandem Heck/decarboxylation/Heck strategy: protecting-group-free synthesis of symmetric and unsymmetric hydroxylated stilbenoids.
PMID 23124823
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
409464
Zinc acrylate, 98%
C6H6O4Zn