EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International Edition in English)
FeCl3-mediated Friedel-Crafts hydroarylation with electrophilic N-acetyl indoles for the synthesis of benzofuroindolines.
PMID 23125000