EMAIL THIS PAGE TO A FRIEND
Cardiology
Ischemic preconditioning of the right ventricle: a new application for an old concept?.
PMID 23128848
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H135
5-Hydroxydecanoic acid sodium salt, ≥97% (HPLC)
C10H19NaO3