EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Synthesis and fluorescence study of a quaternized copolymer containing pyrene for DNA-hybridization detection.
PMID 23129170
Abstract
Built-in detector: A quaternized copolymer containing pyrene is described, which can bind to single-stranded DNA by electrostatic interactions to form a novel complex probe for the detection of DNA hybridization through strengthened fluorescence quenching through its intercalation into double-stranded DNA (see figure).