EMAIL THIS PAGE TO A FRIEND
Breast cancer research and treatment
Plasma estrone sulfate concentrations and genetic variation at the CYP19A1 locus in postmenopausal women with early breast cancer treated with letrozole.
PMID 23129173
Abstract
Estrogen synthesis suppression induced by aromatase inhibitors in breast cancer (BC) patients may be affected by single nucleotide polymorphisms (SNPs) of the gene encoding aromatase enzyme, CYP19A1. We assessed the association between plasma estrone sulfate (ES), letrozole treatment, and four SNPs of CYP19A1 gene (rs10046 C>T, rs4646 G>T, rs749292 C>T, rs727479 T>G) which seem to be related to circulating estrogen levels. Patients were enrolled into a prospective, Italian multi-center clinical trial (Gruppo Italiano Mammella, GIM-5) testing the association of CYP19A1 SNPs with the efficacy of letrozole adjuvant therapy, in postmenopausal early BC patients. SNPs were identified from peripheral blood cell DNA. Plasma ES concentrations were evaluated by Radio Immuno Assay. Blood samples were obtained immediately before letrozole therapy (N = 204), at 6-weeks (N = 178), 6 (N = 152) and 12-months (N = 136) during treatment. Medians (IQR) of ES were 160 pg/mL (85-274) at baseline, 35 pg/mL (12-64) at 6-weeks, 29 pg/mL (17-48) at 6 months and 25 pg/mL (8-46) after 12 months treatment. No statistically significant association was evident between polymorphisms and ES circulating levels during letrozole therapy. Letrozole suppression of the aromatase enzyme function is not affected by polymorphisms of CYP19A1 gene in postmenopausal BC patients.
Related Materials