EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
Decreased entry of calcium into motor nerve endings upon activation of presynaptic cholinergic receptors.
PMID 23129273