EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Novel xylanase from a holstein cattle rumen metagenomic library and its application in xylooligosaccharide and ferulic Acid production from wheat straw.
PMID 23134352
Abstract
A novel gene fragment containing a xylanase was identified from a Holstein cattle rumen metagenomic library. The novel xylanase (Xyln-SH1) belonged to the glycoside hydrolase family 10 (GH10) and exhibited a maximum of 44% identity to the glycoside hydrolase from Clostridium thermocellum ATCC 27405. Xyln-SH1 was heterologously expressed, purified, and characterized. A high level of activity was obtained under the optimum conditions of pH 6.5 and 40 °C. A substrate utilization study indicated that Xyln-SH1 was cellulase-free and strictly specific to xylan from softwood. The synergistic effects of Xyln-SH1 and feruloyl esterase (FAE-SH1) were observed for the release of xylooligosaccharides (XOS) and ferulic acid (FA) from wheat straw. In addition, a high dose of Xyln-SH1 alone was observed to improve the release of FA from wheat straw. These features suggest that this enzyme has substantial potential to improve biomass degradation and industrial applications.
Related Materials