EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Regulating compounding pharmacies after NECC.
PMID 23134357
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1755000 Methylprednisolone acetate
C24H32O6