EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Electrophilic reactivity of tetrabromorhodamine 123 is bromine induced: convergent interpretation through complementary molecular descriptors.
PMID 23134615
Abstract
Nucleophilic addition of water and of methanol to 3,6-diamino-2,4,5,7-tetrabromo-9-[2-(methoxycarbonyl) phenyl]-9H-xanthen-9-ylium, 4BrR123, yields respectively 2-(3,6-diamino-2,4,5,7-tetrabromo-9-hydroxy-9H-xanthen-9-yl)xanthyl benzoate, HO4BrR123 and 2-(3,6-diamino-2,4,5,7-tetrabromo-9-methoxy-9H-xanthen-9-yl)xanthyl benzoate, MeO4BrR123. The novel experimental results are addressed theoretically. The linear free energy relationship, LFER, second-order perturbation theory analysis of the natural bond orbital, NBO, and quantum theory of atoms in molecules, QTAIM, lead to the same conclusion: the electron-withdrawing effect of bonded Br atoms in 4BrR123 extremely enhances the molecular electrophilicity, as compared to 3,6-diamino-9-[2-(methoxycarbonyl) phenyl]-9H-xanthen-9-ylium, R123. The reactivity of these diaminoxanthylium cations is discussed in the context of local and global softness in extended conjugated systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277576
Bromine, ACS reagent, ≥99.5%
Br2
207888
Bromine, reagent grade
Br2