EMAIL THIS PAGE TO A FRIEND
Immunopharmacology and immunotoxicology
Amygdalin induces apoptosis in human cervical cancer cell line HeLa cells.
PMID 23137229
Abstract
Amygdalin, a naturally occurring substance, has been suggested to be efficacious as an anticancer substance. The effect of amygdalin on cervical cancer cells has never been studied. In this study, we found that the viability of human cervical cancer HeLa cell line was significantly inhibited by amygdalin. 4,6-Diamino-2-phenyl indole (DAPI) staining showed that amygdalin-treated HeLa cells developed typical apoptotic changes. The development of apoptosis in the amygdalin-treated HeLa cells were confirmed by double staining of amygdalin-treated HeLa cells with annexin V-FITC and propidium iodide (PI) along with increase in caspase-3 activity in these cells. Further studies indicated that antiapoptotic protein Bcl-2 was downregulated whereas proapoptotic Bax protein was upregulated in the amygdalin-treated HeLa cells implying involvement of the intrinsic pathway of apoptosis. In vivo, amygdalin administration inhibited the growth of HeLa cell xenografts through a mechanism of apoptosis. The results in the present study suggest that amygdalin may offer a new therapeutic option for patients with cervical cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6005
Amygdalin, from apricot kernels, ≥99%
C20H27NO11
10050
Amygdalin, BioXtra, ≥97.0% (HPLC)
C20H27NO11