EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Overcoming the solvation shell during the crystallisation of diatrizoic acid from dimethylsulfoxide.
PMID 23139933
Abstract
We present three crystal structures of diatrizoic acid (DTA) DMSO solvates illustrating progressive desolvation of the DTA molecule and concomitant increasing degree of DTA self-association. Using these structures, plausible pathways for overcoming the solvation shell of DTA during its crystallisation from solution are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9268
Diatrizoic acid
C11H9I3N2O4