EMAIL THIS PAGE TO A FRIEND
Zhonghua kou qiang yi xue za zhi = Zhonghua kouqiang yixue zazhi = Chinese journal of stomatology
[Ultraviolet radiation can improve the biological activity of titanium surface].
PMID 23141665
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF03980932
Titanium, mesh, 150x150mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
GF16031843
Titanium, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
GF11328252
Titanium, mesh, 50x50mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Ti
GF71783354
Titanium, wire reel, 100m, diameter 0.75mm, as drawn, 99.6+%
Ti
GF76735963
Titanium, wire reel, 10m, diameter 0.75mm, as drawn, 99.6+%
Ti
GF29374005
Titanium, mesh, 100x100mm, nominal aperture 4.3mm, wire diameter 1.5mm, 4.4x4.4 wires/inch, open area 94%, platinized diamond mesh
Ti
GF34739837
Titanium, sphere, 200pcs, diameter 3mm, 99.6+%
Ti
GF64061760
Titanium, rod, 1000mm, diameter 16mm, annealed, 99.6+%
Ti
GF26694771
Titanium, wire reel, 20m, diameter 0.125mm, annealed, 99.6+%
Ti
GF70333376
Titanium, wire reel, 10m, diameter 0.05mm, hard, 99.8%
Ti
GF96834493
Titanium, rod, 1000mm, diameter 5mm, annealed, 99.6+%
Ti
GF16213553
Titanium, wire reel, 10m, diameter 0.125mm, annealed, 99.6+%
Ti
GF76266423
Titanium, wire reel, 0.5m, diameter 1.0mm, annealed, 99.6+%
Ti
GF46132534
Titanium, wire reel, 100m, diameter 0.25mm, as drawn, 99.6+%
Ti
GF76772391
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Ti
GF98611623
Titanium, rod, 200mm, diameter 8.0mm, as drawn, 99.99+%
Ti
GF66991179
Titanium, rod, 100mm, diameter 2mm, as drawn, 99.99+%
Ti
GF58567067
Titanium, rod, 100mm, diameter 5mm, annealed, 99.6+%
Ti
GF33043069
Titanium, rod, 100mm, diameter 30mm, annealed, 99.6+%
Ti
GF34996685
Titanium, wire reel, 200m, diameter 0.125mm, annealed, 99.6+%
Ti
GF13900205
Titanium, tube, 100mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Ti
GF71724124
Titanium, tube, 500mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Ti
GF50643479
Titanium, wire reel, 5m, diameter 1.5mm, as drawn, 99.6+%
Ti
GF90688449
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Ti
GF51690663
Titanium, wire reel, 5m, diameter 0.5mm, as drawn, 99.6+%
Ti
GF00558187
Titanium, wire reel, 25m, diameter 1.0mm, as drawn, 99.6+%
Ti
GF39040084
Titanium, rod, 100mm, diameter 16mm, annealed, 99.6+%
Ti
GF81436057
Titanium, rod, 500mm, diameter 4mm, annealed, 99.6+%
Ti
GF62263841
Titanium, wire reel, 50m, diameter 0.125mm, annealed, 99.6+%
Ti
GF61322699
Titanium, rod, 100mm, diameter 2mm, annealed, 99.6+%
Ti