EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Propargyl hydrazides: synthesis and conversion into pyrazoles through hydroamination.
PMID 23143924
Abstract
Pyrazoles direct: propargyl alcohols undergo hydrazination when treated with p-tosyl hydrazide in the presence of catalytic amounts of either Sc(OTf)(3) or La(OTf)(3) (see scheme; Tf=trifluoromethanesulfonyl). Propargyl hydrazides are converted into either N-tosyl or N-H pyrazoles when treated with an acid or a base, respectively. The one-step acid-catalyzed hydrazination/cyclization of propargyl alcohols directly affords pyrazoles in high yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50803
Propargyl alcohol, 99%
C3H4O