EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Mechanistic understanding of polycyclic aromatic hydrocarbons (PAHs) from the thermal degradation of tires under various oxygen concentration atmospheres.
PMID 23145576
Abstract
The thermal degradation of tires under various oxygen concentrations (7-30%/Bal. N(2)) was investigated thermo-gravimetrically at 10 °C min(-1) heating rate over a temperature range from ambient to 1000 °C. Significant mass loss (~55%) was observed at the temperature of 300-500 °C, where the thermal degradation rate was almost identical and independent of oxygen concentrations due to simultaneous volatilization and oxidation. A series of gas chromatography/mass spectroscopy (GC/MS) measurements taken from the effluent of a thermo-gravimetric analysis (TGA) unit at temperature of 300-5000 °C leads to the overall thermal degradation mechanisms of waste tires and some insights for understanding evolution steps of air pollutants including volatile organic carbons (VOCs) and polycyclic aromatic hydrocarbons (PAHs). In order to describe the fundamental mechanistic behavior on tire combustion, the main constituents of tires, styrene butadiene rubber (SBR) and polyisoprene (IR), has been investigated in the same experimental conditions. The thermal degradation of SBR and IR suggests the reaction mechanisms including bond scissions followed by hydrogenation, gas phase addition reaction, and/or partial oxidation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
651796
Negative photoresist I