EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Ras-guanine-nucleotide-releasing factors 1 and 2 interact with PLCγ at focal adhesions to enable IL-1-induced Ca(2+) signalling, ERK activation and MMP-3 expression.
PMID 23145787
Abstract
IL (interleukin)-1 signalling in anchorage-dependent cells involves focal-adhesion-restricted and Ca2+-dependent Ras and ERK (extracellular-signal-regulated kinase) activation that leads to MMP (matrix metalloproteinase) release and extracellular matrix remodelling. Ras activity is regulated, in part, by the Ca2+-responsive Ras GRFs (guanine-nucleotide-releasing factors) 1 and 2, but the mechanisms that link and localize IL-1-induced Ca2+ signalling to focal adhesions are not defined. In the present study we characterized the role of Ras-GRF1/2 in Ca2+ and Ras→ERK signalling after IL-1 stimulation. By immunoprecipitation we found that Ras-GRF1/2 associates with PLCγ1 (phospholipase Cγ1). This association enables PLCγ1 recruitment to focal adhesions and is required for Ras signalling, ERK activation and MMP-3 release downstream of IL-1 stimulation. Depletion of PLCγ1 by siRNA (small interfering RNA) abolished IL-1-induced Ras activation and MMP-3 expression. Buffering of cytosolic Ca2+ reduced Ras interactions with Ras-GRF1/2 and blocked MMP-3 release. The results of the present study show that, in addition to their functions as Ras-exchange factors, Ras-GRF1 and -GRF2 may act as adaptors that bind PLCγ1 and restrict Ca2+ signalling to the vicinity of focal adhesions, indicating a new role for these GRFs that is required for IL-1 induction of the Ras→ERK pathway and MMP-3 expression.