EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Effects of St John's wort and its active constituents, hypericin and hyperforin, on isolated rat urinary bladder.
PMID 23146040
Abstract
To investigate the effect of St John's wort (SJW) and its active constituents hypericin and hyperforin on detrusor smooth muscle contractility and their possible neuroprotective role against ischaemic-like conditions, which could arise during overactive bladder disease. In whole bladders, intrinsic nerves underwent electrical field stimulation (EFS). The effect of drugs on the contractile response and its recovery in reperfusion phase (R) was monitored at different concentrations during 1 or 2 h of anoxia-glucopenia (A-G) and the first 30 min of R. The effects of the drugs were also investigated on rat detrusor muscle strips contracted with carbachol, KCl and electrically. SJW has spasmolytic activity, which increases with increasing concentration and it worsens the damage induced by A-G/R on rat urinary bladder. Hypericin and hyperforin had no effect during ischemic-like conditions but they both exert a dual modulation of rat detrusor strips contraction. At high micromolar concentrations they showed a relaxing effect, but at submicromolar range hypericin increased the plasma membrane depolarisation and hyperforin showed a stimulatory effect on the cholinergic system. The results of our study showed that SJW and its constituents could modulate urinary bladder contractility and even worsen A-G/R injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00190585
Hypericin, primary reference standard
C30H16O8