EMAIL THIS PAGE TO A FRIEND
Diabetes research and clinical practice
Seven-day subcutaneous continuous glucose monitoring demonstrates that treatment with acarbose attenuates late dumping syndrome in a woman with gastrectomy for gastric cancer.
PMID 23146372