EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Quinalphos induced oxidative stress and histoarcheitectural alterations in adult male albino rats.
PMID 23146591
Abstract
Quinalphos is a synthetic organophosphate used as a broad spectrum insecticide and acaricide. The present study investigates the effect of three sub-lethal doses (0.52, 1.04, 2.6 mg/kg b.wt) of quinalphos for variable durations (15, 30 and 90 days) on oxidative stress and histopathological changes in adult male rats. Quinalphos treatment for 15 and 30 days resulted in a dose dependent significant increase in malondialdehyde (MDA) levels and glutathione-S-transferase (GST) activity together with a concurrent decrease in ferric reducing ability of plasma (FRAP) and glutathione (GSH) content. Quinalphos treatment for 90 days also induced a significant increase in MDA levels and GST activity but the effect was not dose-dependent. Histopathological examination of liver revealed architectural disarray and dilatation of sinusoids, focal fatty changes, accumulation of eosinophils and single cell necrosis with increasing doses. However, spleen and kidney did not show any histological changes. Administration of quinalphos resulted in oxidative stress and free radical induced injury as evidenced by increased lipid peroxidation, decreased FRAP and histopathological changes in liver.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45651
Quinalphos, PESTANAL®, analytical standard
C12H15N2O3PS