EMAIL THIS PAGE TO A FRIEND
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology
Psychoactive 'bath salts': compounds, mechanisms, and toxicities.
PMID 23147485