EMAIL THIS PAGE TO A FRIEND
Organic letters
Asymmetric iodolactonization utilizing chiral squaramides.
PMID 23148494
Abstract
Asymmetric iodolactonization of γ- and δ-unsaturated carboxylic acids has been explored in the presence of six different chiral organocatalysts 5-8. The catalyst 6b was found to facilitate the cyclization of 5-arylhex-5-enoic acids 1 to the corresponding iodolactones 2 with up to 96% ee. By this protocol, unsaturated carboxylic acids are converted enantioselectively to synthetically useful δ-lactones in high yields using commercially available NIS. Apparently, both hydrogen bonding and aryl/aryl interactions are important for efficient stereodifferentiation.