EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Design, synthesis, and evaluation of (E)-N-substituted benzylidene-aniline derivatives as tyrosinase inhibitors.
PMID 23149255
Abstract
We attempted to design and synthesize (E)-N-substituted benzylidene-hydroxy or methoxy-aniline derivatives and to evaluate their inhibitory effect on tyrosinase activity and anti-melanogenesis activity in murine B16F10 melanoma cells. Derivatives with a 4-methoxy- or 4-hydroxy-anilino group exerted more potent inhibition against mushroom tyrosinase than those with a 2-hydroxyanilino group. (E)-4-((4-Hydroxyphenylimino)methyl)benzene-1,2-diol exhibited the most potent and non-competitive inhibition on mushroom tyrosinase showing an IC(50) of 17.22 ± 0.38 μM and being more effective than kojic acid (51.11 ± 1.42 μM). This compound decreased melanin production stimulated by the alpha-melanocyte-stimulating hormone and inhibited murine tyrosinase activity in a dose-dependent manner. Therefore, we propose (E)-4-((4-hydroxyphenylimino)methyl)benzene-1,2-diol as a new candidate of potent tyrosinase inhibitors that could be used as therapeutic agent with safe skin-whitening efficiency.