EMAIL THIS PAGE TO A FRIEND
Toxicon : official journal of the International Society on Toxinology
Influence of endophyte genotype on swainsonine concentrations in Oxytropis sericea.
PMID 23149419
Abstract
Locoism is a toxic syndrome of livestock caused by the ingestion of a subset of legumes belonging to the Astragalus and Oxytropis genera known as "locoweeds". Locoweeds contain the toxic indolizidine alkaloid swainsonine, which is produced by the endophytic fungi Undifilum species. Previously we reported that swainsonine concentrations differ between populations of Oxytropis sericea. We hypothesized that the genotype of the plant, endophyte, or an interaction of the two may be responsible for the differences in swainsonine concentration between populations of O. sericea. To test this hypothesis, plants derived from seeds collected at each location were grown in a common garden, Undifilum oxytropis isolates from each location were cultured and grown in a common environment, and a plant genotype by endophyte cross inoculation was performed. Here we show that the genotype of the endophyte is responsible for the differences in swainsonine concentrations observed in the two populations of O. sericea.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9263
Swainsonine, synthetic
C8H15NO3
S8195
Swainsonine, from Metarrhizium anisopliae, ≥98% (TLC)
C8H15NO3