EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Statins inhibit in vitro virulence phenotypes of Pseudomonas aeruginosa.
PMID 23149514
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2537
Mevastatin, ≥95% (HPLC), powder
C23H34O5