EMAIL THIS PAGE TO A FRIEND
Giornale italiano di dermatologia e venereologia : organo ufficiale, Societa italiana di dermatologia e sifilografia
Consensus on the use of the fixed combination calcipotriol/betamethasone dipropionate in the treatment of plaque psoriasis.
PMID 23149707
Abstract
Calcipotriol, a vitamin D analogue, and betamethasone dipropionate, a high potency corticosteroid, are complementary agents for the topical treatment of psoriasis vulgaris. Robust evidence on the efficacy and safety of their fixed combination has been provided by randomized, double-blind, controlled clinical trials involving more than 7000 patients with the ointment formulation in psoriasis of the body and more than 4000 patients with the gel formulation in scalp psoriasis. These trials have shown that the fixed combination ointment is more effective and better tolerated, not only than placebo, but also than calcipotriol and tacalcitol monotherapies. In addition, it has proved, in most instances, to be more effective than betamethasone and at least as well tolerated. The same applies to the gel for scalp and body psoriasis. Safety studies have excluded that repeated courses of treatment with the fixed combination for up to one year produce systemic effects. Studies have also shown that the fixed combination treatment improves quality of life to a significantly greater extent than calcipotriol, with the once daily regimen most appreciated by patients, in both active disease and recurrency. Because of the extensive evidence, American and European guidelines recommend the calcipotriol/betamethasone dipropionate fixed combination as first line topical treatment for mild to moderate plaque psoriasis of the body and scalp.