EMAIL THIS PAGE TO A FRIEND
Cancer Research
Bcl3 selectively promotes metastasis of ERBB2-driven mammary tumors.
PMID 23149915
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-697
Anti-E-Cadherin Antibody