EMAIL THIS PAGE TO A FRIEND
Parasites & vectors
Optimizing molluscicide treatment strategies in different control stages of schistosomiasis in the People's Republic of China.
PMID 23151396
Abstract
The application of chemical molluscicides is still one of the most effective measures for schistosomiasis control in P. R. China. By applying diverse molluscicide treatment scenarios on different snail densities in the field, we attempted to understand the cost-effectiveness of molluscicide application so as to prescribe an optimal management approach to control intermediate host snail Oncomelania hupensis under acceptable thresholds based on the goal of the National Schistosomiasis Control Programme. The molluscicidal field trial was carried out in the marshland of an island along the Yangtze River, Jiangsu province, P.R. China in October 2010. Three plots in the island representing low-density, medium-density and high-density groups were identified after the baseline survey on snail density. Each snail density plot was divided into four experimental units in which molluscicide (50% niclosamide ethanolamine salt wettable powder) was applied once, twice, trice and four times, respectively. The logistic regression model to correlate snail mortality rate with the covariates of number of molluscicidal treatment and snail density, and a linear regression model to investigate the relationship between cost-effectiveness and number of molluscicidal treatment as well as snail density were established. The study revealed that increase in the number of molluscicide treatments led to increased snail mortality across all three population density groups. The most cost-effective regimen was seen in the high snail density group with a single molluscicide treatment. For both high and low density groups, the more times molluscicide were applied, the less cost-effectiveness was. However, for the median density group, the level of cost-effectiveness for two applications was slightly higher than that in one time. We concluded that different stages of the national schistosomiasis control/elimination programme, namely morbidity control, transmission control and transmission interruption, should utilize different molluscicide treatment strategies to maximize cost-effectiveness.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3510
Niclosamide
C13H8Cl2N2O4
N0560000
Niclosamide (anhydrous), European Pharmacopoeia (EP) Reference Standard
C13H8Cl2N2O4