EMAIL THIS PAGE TO A FRIEND
The Cochrane database of systematic reviews
Interventions for preventing recurrent urinary tract infection during pregnancy.
PMID 23152271
Abstract
Recurrent urinary tract infections (RUTI) are common in women who are pregnant and may cause serious adverse pregnancy outcomes for both mother and child including preterm birth and small-for-gestational-age babies. Interventions used to prevent RUTI in women who are pregnant can be pharmacological (antibiotics) or non-pharmacological (cranberry products, acupuncture, probiotics and behavioural modifications). So far little is known about the best way to prevent RUTI in pregnant women. To assess the effects of interventions for preventing recurrent urinary tract infections in pregnant women.The primary maternal outcomes were RUTI before birth (variously defined) and preterm birth (before 37 weeks). The primary infant outcomes were small-for-gestational age and total mortality. We searched the Cochrane Pregnancy and Childbirth Group's Trials Register (8 June 2012) and reference lists of retrieved articles. Published, unpublished and ongoing randomised controlled trials (RCTs), quasi-RCTs, clustered-randomised trials and abstracts of any intervention (pharmacological and non-pharmacological) for preventing RUTI during pregnancy (compared with another intervention, placebo or with usual care). Two review authors independently evaluated the one identified trial for inclusion and assessed trial quality. Two review authors extracted data. Data were checked for accuracy. The review included one trial involving 200 women. The trial compared a daily dose of nitrofurantoin and close surveillance (regular clinic visit, urine cultures and antibiotics when a positive culture was found) with close surveillance only. No significant differences were found for the primary outcomes: recurrent pyelonephritis (risk ratio (RR) 0.89, 95% confidence interval (CI) 0.31 to 2.53, one study, 167 women), recurrent urinary tract infection before birth (RR 0.30, 95% CI 0.06 to 1.38; one study 167 women) and preterm birth (before 37 weeks) (RR 1.18, 95% CI 0.42 to 3.35; one study 147 women). The incidence of asymptomatic bacteriuria (ASB) (at least 10(3) colonies per mL) (secondary outcome), only reported in women with a clinic attendance rate of more than 90% (RR 0.55, 95% CI 0.34 to 0.89; one study, 102 women), was significantly reduced in women who received nitrofurantoin and close surveillance. A daily dose of nitrofurantoin and close surveillance has not been shown to prevent RUTI compared with close surveillance alone. A significant reduction of ASB was found in women with a high clinic attendance rate and who received nitrofurantoin and close surveillance. There was limited reporting of both primary and secondary outcomes for both women and infants. No conclusions can be drawn regarding the optimal intervention to prevent RUTI in women who are pregnant. Randomised controlled trials comparing different pharmacological and non-pharmacological interventions are necessary to investigate potentially effective interventions to prevent RUTI in women who are pregnant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1191
Nitrofurantoin, Pharmaceutical Secondary Standard; Certified Reference Material
C8H6N4O5