EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Hesperetin: an inhibitor of the transforming growth factor-β (TGF-β) signaling pathway.
PMID 23153811
Abstract
We have identified a previously unknown function of the natural compound, hesperetin. Here, we demonstrate that this small molecular agent is able to inhibit the transforming growth factor-β (TGF-β) signaling pathway. Single-molecule force measurements and single-molecule fluorescence imaging show that hesperetin interferes with ligand-receptor interactions. Furthermore, by Western blot analysis, it was confirmed that hesperetin also inhibits the phosphorylation of Smad3, a down-stream target of the TGF-β pathway. In addition we demonstrated that this compound hinders TGF-β1-induced cancer cell migration and invasion. These results suggest a potential future application for hesperetin as a TGF-β inhibitor in cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50162
Hesperidin, analytical standard
C28H34O15