EMAIL THIS PAGE TO A FRIEND
Hypertension research : official journal of the Japanese Society of Hypertension
Effects of valsartan on progression of kidney disease in Japanese hypertensive patients with advanced, predialysis, chronic kidney disease: Kanagawa Valsartan Trial (KVT).
PMID 23154591
Abstract
Suppression of the renin-angiotensin system is known to slow progression of chronic kidney disease (CKD). However, few trials have been performed with Japanese patients. This study investigated whether the angiotensin receptor blocker (ARB) valsartan would delay the progression of kidney disease more effectively than conventional treatment in Japanese hypertensive patients with advanced, predialysis CKD. In a multicenter, randomized, open-label trial, 303 patients with hypertension and CKD with serum creatinine levels 2.0 mg dl(-1) were assigned to receive either conventional therapy plus valsartan (valsartan add-on group) or conventional therapy without ARB (control group). The primary outcome was a change in serum creatinine levels. Changes in urinary protein levels and time to onset of renal events were analyzed as secondary end points. There were no between-group differences in blood pressure during the study. Changes in serum creatinine and urinary protein levels did not differ between the groups. However, the rate of renal events, including doubling of serum creatinine levels or end-stage renal disease, was significantly lower in the valsartan add-on group than in the control group. The addition of valsartan decreased the risk by 42.6% after adjustment for baseline variables. The addition of valsartan to conventional therapy significantly slowed the rate of renal function decline and delayed the need for renal replacement therapy in Japanese hypertensive patients with advanced CKD.