EMAIL THIS PAGE TO A FRIEND
Internal medicine (Tokyo, Japan)
ANCA-associated vasculitis with central retinal artery occlusion developing during treatment with methimazole.
PMID 23154728
Abstract
A 63-year-old woman suddenly developed central retinal artery occlusion following a slight fever while being treated with methimazole (MMI) for hyperthyroidism. She was diagnosed to have anti-neutrophil cytoplasmic antibody (ANCA)-associated vasculitis (AAV) based on increased inflammatory reactions with positive myeloperoxidase-ANCA in the serum. Her visual acuity remained low despite immediate treatment with corticosteroids and cyclophosphamide after cessation of MMI, which may have played a role in the pathogenesis of AAV. Central retinal artery occlusion is a rare manifestation of AAV; however, it is important with regard to the possibility of serious sequelae.