EMAIL THIS PAGE TO A FRIEND
Journal of applied physiology (Bethesda, Md. : 1985)
Vasoconstrictor responsiveness during hyperbaric hyperoxia in contracting human muscle.
PMID 23154993
Abstract
Large increases in systemic oxygen content cause substantial reductions in exercising forearm blood flow (FBF) due to increased vascular resistance. We hypothesized that 1) functional sympatholysis (blunting of sympathetic α-adrenergic vasoconstriction) would be attenuated during hyperoxic exercise and 2) α-adrenergic blockade would limit vasoconstriction during hyperoxia and increase FBF to levels observed under normoxic conditions. Nine male subjects (age 28 ± 1 yr) performed forearm exercise (20% of maximum) under normoxic and hyperoxic conditions. Studies were performed in a hyperbaric chamber at 1 atmosphere absolute (ATA; sea level) while breathing 21% O(2) and at 2.82 ATA while breathing 100% O(2) (estimated change in arterial O(2) content ∼6 ml O(2)/100 ml). FBF (ml/min) was measured using Doppler ultrasound. Forearm vascular conductance (FVC) was calculated from FBF and blood pressure (arterial catheter). Vasoconstrictor responsiveness was determined using intra-arterial tyramine. FBF and FVC were substantially lower during hyperoxic exercise than normoxic exercise (∼20-25%; P < 0.01). At rest, vasoconstriction to tyramine (% decrease from pretyramine values) did not differ between normoxia and hyperoxia (P > 0.05). During exercise, vasoconstrictor responsiveness was slightly greater during hyperoxia than normoxia (-22 ± 3 vs. -17 ± 2%; P < 0.05). However, during α-adrenergic blockade, hyperoxic exercise FBF and FVC remained lower than during normoxia (P < 0.01). Therefore, our data suggest that although the vasoconstrictor responsiveness during hyperoxic exercise was slightly greater, it likely does not explain the majority of the large reductions in FBF and FVC (∼20-25%) during hyperbaric hyperoxic exercise.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7547
Phentolamine hydrochloride, ≥98% (TLC), powder
C17H19N3O · HCl
P7561
Phentolamine methanesulfonate salt, ≥98% (TLC), powder
C17H19N3O · CH4O3S