EMAIL THIS PAGE TO A FRIEND
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic]
[Rapid analysis of suppositories by quantitative 1H NMR spectroscopy].
PMID 23156037
Abstract
Rapid analysis of suppositories with ibuprofen and arbidol by quantitative 1H NMR spectroscopy was performed. Optimal conditions for the analysis were developed. The results are useful for design of rapid methods for quality control of suppositories with different components
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0860 Arbidol hydrochloride, ≥98% (HPLC)
C22H25BrN2O3S · HCl