EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Antihypertensive and neuroprotective activity of quercetin and its derivatives].
PMID 23156087
Abstract
Data available in the literature on the pharmacological activity of quercetin and its derivatives, which possess various biological properties including antihypertensive and neurotropic activity, are reviewed. The mechanisms of these effects and results of clinical trials are considered and prospects for the clinical use of quercetin and its derivatives are discussed.