EMAIL THIS PAGE TO A FRIEND
Nihon rinsho. Japanese journal of clinical medicine
[Adverse effects of cardiovascular agents in Japan--update 2012].
PMID 23156515
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7455
Tolvaptan, ≥98% (HPLC), powder
C26H25ClN2O3