EMAIL THIS PAGE TO A FRIEND
The European Journal of Neuroscience
Influence of chronic amphetamine treatment and acute withdrawal on serotonin synthesis and clearance mechanisms in the rat ventral hippocampus.
PMID 23157166
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB1501R
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse