EMAIL THIS PAGE TO A FRIEND
Zhonghua bing li xue za zhi = Chinese journal of pathology
[Thallium poisoning: report of an autopsy case].
PMID 23157754
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277932
Thallium, granular, 1-5 mm diameter, 99.99% trace metals basis
Tl