EMAIL THIS PAGE TO A FRIEND
Talanta
Development and validation of a simple and sensitive high performance liquid chromatographic method for the simultaneous determination of anastrozole, bicalutamide, tamoxifen, and their synthetic impurities.
PMID 23158354
Abstract
A simple and sensitive analytical method for simultaneous determination of anastrozole, bicalutamide, and tamoxifen as well as their synthetic impurities, anastrozole pentamethyl, bicalutamide 3-fluoro-isomer, and tamoxifen e-isomer, was developed and validated by using high performance liquid chromatography (HPLC). The separation was achieved on a Symmetry(®) C-8 column (100×4.6 mm i.d., 3.5 μm) at room temperature (±24 °C), with a mobile phase consisting of acetonitrile/water containing 0.18% N,N dimethyloctylamine and pH adjusted to 3.0 with orthophosphoric acid (46.5/53.5, v/v) at a flow rate of 1.0 mL min(-1) within 20 min. The detection was made at a wavelength of 270 nm by using ultraviolet (UV) detector. No interference peaks from excipients and relative retention time indicated the specificity of the method. The calibration curve showed correlation coefficients (r) >0.99 calculated by linear regression and analysis of variance (ANOVA). The limit of detection (LOD) and limit of quantitation (LOQ), respectively, were 2.2 and 6.7 μg mL(-1) for anastrozole, 2.61 and 8.72 μg mL(-1) for bicalutamide, 2.0 and 6.7 μg mL(-1) for tamoxifen, 0.06 and 0.22 μg mL(-1) for anastrozole pentamethyl, 0.02 and 0.07 μg mL(-1) for bicalutamide 3-fluoro-isomer, and 0.002 and 0.007 μg mL(-1) for tamoxifen e-isomer. Intraday and interday relative standard deviations (RSDs) were <2.0% (drugs) and <10% (degradation products) as well as the comparison between two different analysts, which were calculated by f test.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2736
Anastrozole, ≥98% (HPLC)
C17H19N5
B9061
Bicalutamide (CDX), ≥98% (HPLC), powder
C18H14F4N2O4S