EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Oral cholestyramine is not an effective treatment for uncomplicated erythropoietic protoporphyria.
PMID 23158617
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4650 Cholestyramine resin