EMAIL THIS PAGE TO A FRIEND
Zhonghua zhong liu za zhi [Chinese journal of oncology]
[Clinical analysis of childhood and adolescent Hodgkin's lymphoma: a report of 88 cases].
PMID 23159084
Abstract
The aim of this study was to investigate the clinicopathological characteristics, effective treatment and prognosis in childhood and adolescent Hodgkin's lymphoma. A total of 88 patients with childhood and adolescent Hodgkin's lymphoma were treated in the Cancer Hospital of CAMS from 1998 to 2005. The clinicopathological and follow-up data of the patients were retrospectively reviewed. The survival rate was calculated by Kaplan-Meier method and compared by log-rank test. COX multivariate prognosis analysis was performed. The 2-year event-free survival rate of the 88 patients was 86.4%, the 5-year event-free survival rate was 61.4%, and the 5-year overall survival rate was 95.5%. Univariate analysis showed that the stage of disease (P = 0.033), "B" symptoms (P = 0.028), bulky disease (P = 0.007), splenomegaly (P = 0.050), LDH elevation (P = 0.020), chemotherapy regimen (P = 0.003) were prognostic factors in the 5-year event-free survival rate. Splenomegaly (P = 0.039), LDH elevation (P = 0.033), chemotherapy regimen (P = 0.008) were prognostic factors of 5-year overall survival rate. Multivariate analysis showed that chemotherapy regimen (P = 0.033), stage of disease (P = 0.023), LDH elevation (P = 0.008), "B" symptoms (P = 0.044), bulky disease (P = 0.009) were independent prognostic factors of 5-year event-free survival rate. The chemotherapy regimen (P = 0.012) and LDH elevation (P = 0.046) were independent prognostic factors of 5-year overall survival rate. The non-ABVD chemotherapy regimen, stage IV disease, LDH elevation, associated with "B" symptoms and bulky disease are independent prognostic factors of 5-year event-free survival rate. LDH elevation and non-ABVD chemotherapy regimen are independent prognostic factors of 5-year overall survival rate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0036
Procarbazine hydrochloride, ≥98% (HPLC)
C12H19N3O·HCl