EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Reduction of borazines mediated by low-valent chromium species.
PMID 23161604
Abstract
Recharging spent BN fuel: {Cr(CO)(3)} mediates the reduction of borazines by hydride and methyl nucleophiles to generate anionic complexes of dearomatized hexamethylborazine. Subsequent quenching leads to the release of a substituted cyclotriborazane, successfully demonstrating the stepwise reduction of a B=N bond.