EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Synthesis of highly functionalized fluorinated cispentacin derivatives.
PMID 23161635
Abstract
Fluorinated highly functionalized cispentacin derivatives were synthetised starting from an unsaturated bicyclic β-lactam through C=C bond functionalization via the dipolar cycloaddition of a nitrile oxide, isoxazoline opening, and fluorination by OH/F exchange.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A48105
Cycloleucine, 97%
C6H11NO2